missing translation for 'onlineSavingsMsg'
Learn More

Lumefantrine 98.0+%, TCI America™

Supplier:  TCI America L025625G

 View more versions of this product

Catalog No. L025625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lumefantrine
136°C
C30H32Cl3NO
25 g
DYLGFOYVTXJFJP-MYYYXRDXNA-N
2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
6437380
528.94
Crystalline Powder
82186-77-4
Yellow
MFCD05662268
Benflumetol, (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol
CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
528.94
CHEBI:156095
≥98.0% (HPLC,T)

Chemical Identifiers

82186-77-4
528.94
DYLGFOYVTXJFJP-MYYYXRDXNA-N
6437380
2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
C30H32Cl3NO
MFCD05662268
Benflumetol, (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol
CHEBI:156095
CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions