accessibility menu, dialog, popup

UserName

CH3NCH3OHClClCl(Z)

CAS RN 82186-77-4

IUPAC Name: 2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
Synonyms: lumefantrine dl-benflumelol benflumetol coartem 本芴醇
Molecular Weight (g/mol): 528.94
Molecular Formula: C30H32Cl3NO
InChi Key: DYLGFOYVTXJFJP-MYYYXRDXNA-N
SMILES: CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)