missing translation for 'onlineSavingsMsg'
Learn More

Lumefantrine 98.0+%, TCI America™

Catalog No. p-7131896
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02565G 5 g
L025625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02565G Supplier TCI America Supplier No. L02565G
Only null left

Chemical Identifiers

CAS 82186-77-4
Molecular Formula C30H32Cl3NO
Molecular Weight (g/mol) 528.94
MDL Number MFCD05662268
InChI Key DYLGFOYVTXJFJP-MYYYXRDXNA-N
Synonym Benflumetol, (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol
PubChem CID 6437380
ChEBI CHEBI:156095
IUPAC Name 2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
SMILES CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 136°C
Color Yellow
Quantity 5 g
Formula Weight 528.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lumefantrine