SS320 (MC1061 F') Electrocompetent Cells, 12 Rxns (DUOs)Construct larger, more complex phage display libraries with highest efficiency cells. This amber suppressor strain can be used for phage display and protein expression.Perfect for antibody phage display or peptide phage display library creation. SS320 Electrocompetent Cells are a non-amber suppressor strain (sometimes called MC1061F') prepared as highly efficient (4 10^10 cfu/g) electrocompetent cells for phage display library screening. These cells have the highest available transformation efficiency of any strain used for phage display.