missing translation for 'onlineSavingsMsg'
Learn More

PURELL™ LTX-12™ Dispenser Floor Stand

Catalog No. 19044010
Change view
Click to view available options
Mounting:
Freestanding
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
19044010 Freestanding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19044010 Supplier PURELL™ Supplier No. 7028DS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LTX-12™ touch-free dispenser floor stand for hand sanitizer.

  • Touch-free dispenser stand
  • For PURELL™ Hand Sanitizer

Specifications

Product Type Dispenser Stand
Color Black, Gray
Length (Metric) 28.5 cm
Width (Metric) 13.5 cm
Height (Metric) 5.75 cm
Holds Hand Sanitizer
Material Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Front-loading with Opening
For Use With (Equipment) PURELL&Trade; Hand Sanitizer
Show More Show Less