missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149817
Description
LPA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a serine proteinase that inhibits the activity of tissue-type plasminogen activator I. The encoded protein constitutes a substantial portion of lipoprotein(a) and is proteolytically cleaved, resulting in fragments that attach to atherosclerotic lesions and promote thrombogenesis. Elevated plasma levels of this protein are linked to atherosclerosis. Depending on the individual, the encoded protein contains 2-43 copies of kringle-type domains. The allele represented here contains 15 copies of the kringle-type repeats and corresponds to that found in the reference genome sequence.Identifiants chimiques
| LPA | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08519 | |
| 4018 | |
| Recombinant Lipoprotein, a (Lpa) (Asp1719-Arg2038). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| LPA | |
| AK38; antiangiogenic AK38 protein; apo(a); APOA; apolipoprotein(a); lipoprotein(a); lipoprotein, Lp(a); LP; lp(a); LPA | |
| LPA | |
| Primary | |
| Protein A |
Spécifications
| LPA | |
| Polyclonal | |
| Unconjugated | |
| LPA | |
| AK38; antiangiogenic AK38 protein; apo(a); APOA; apolipoprotein(a); lipoprotein(a); lipoprotein, Lp(a); LP; lp(a); LPA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08519 | |
| LPA | |
| Recombinant Lipoprotein, a (Lpa) (Asp1719-Arg2038). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody |