Learn More
Thermo Scientific Chemicals Loxoprofen sodium
Supplier: Thermo Scientific Chemicals 467120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Loxoprofen sodium | |
| 25 g | |
| C15H17NaO3 | |
| BAZQYVYVKYOAGO-UHFFFAOYNA-M | |
| sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate |
| 80382-23-6 | |
| Solubility aq. soln.: soluble. Other solubilities: soluble in chloroform, methanol | |
| 268.29 | |
| O.O.[Na+].CC(C([O-])=O)C1=CC=C(CC2CCCC2=O)C=C1 | |
| 304.32 |
Chemical Identifiers
| 80382-23-6 | |
| C15H17NaO3 |
Safety and Handling
GHS H Statement Toxic if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Store locked up.
GHS Signal Word: Danger
RUO – Research Use Only