missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Loxoprofen sodium

Catalog No. p-7214776
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AC467120050 5 g
AC467120250 25 g
2 options

Catalog No. AC467120050

Supplier: Thermo Scientific Chemicals 467120050

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80382-23-6
Molecular Formula C15H17NaO3

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in chloroform, methanol
Formula Weight 268.29
InChI Key BAZQYVYVKYOAGO-UHFFFAOYNA-M
SMILES O.O.[Na+].CC(C([O-])=O)C1=CC=C(CC2CCCC2=O)C=C1
IUPAC Name sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate
Molecular Weight (g/mol) 304.32
Chemical Name or Material Loxoprofen sodium
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

RUO – Research Use Only