missing translation for 'onlineSavingsMsg'
Learn More

Lopinavir Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013419
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013419 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013419 Supplier Merck Emd Millipore Supplier No. PHR19271G
Lopinavir is an antiretroviral agent and a protease inhibitor indicated for the management of human immunodeficiency virus (HIV) infection. It is a key component of Kaletra, the co-formulation of lopinavir and ritonavir.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 192725-17-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C37H48N4O5
Synonym Lopinavir
Type Certified Reference Material
Quantity 1 g
Formula Weight 628.80
Grade Certified Reference Material