missing translation for 'onlineSavingsMsg'
Learn More

Loganin 98.0+%, TCI America™

Catalog No. L026850MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L026850MG 50 mg
L026810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L026850MG Supplier TCI America Supplier No. L026850MG

Chemical Identifiers

CAS 18524-94-2
Molecular Formula C17H26O10
Molecular Weight (g/mol) 390.385
MDL Number MFCD00075645
InChI Key AMBQHHVBBHTQBF-UOUCRYGSSA-N
PubChem CID 87691
ChEBI CHEBI:15771
IUPAC Name methyl (1S,4aS,6S,7R,7aS)-6-hydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate
SMILES CC1C(CC2C1C(OC=C2C(=O)OC)OC3C(C(C(C(O3)CO)O)O)O)O

Specifications

Melting Point 225°C
Color White
Quantity 50 mg
Formula Weight 390.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Loganin