missing translation for 'onlineSavingsMsg'
Learn More

Lobenzarit Disodium Salt 98.0+%, TCI America™

Catalog No. L027825MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L027825MG 25 mg
L0278100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L027825MG Supplier TCI America Supplier No. L027825MG
Only null left

Chemical Identifiers

CAS 64808-48-6
Molecular Formula C14H8ClNNa2O4
Molecular Weight (g/mol) 335.651
MDL Number MFCD00941422
InChI Key QPJBONAWFAURGB-UHFFFAOYSA-L
Synonym N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate
PubChem CID 47462
IUPAC Name disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 388°C
Color White-Yellow
Quantity 25 mg
Formula Weight 335.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lobenzarit Disodium Salt
RTECSNumber DG4975520
TSCA No