Learn More
Invitrogen™ LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)
Description
LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)
- A two-color fluorescent cell viability assay that simultaneously identifies live and dead cells
- Live cells are labeled with calcein dye and emit a green fluorescence
- Dead cells are labeled with SYTOX Deep Red dye and emit a red fluorescence
- Dyes can be multiplexed with other orange/red fluorophores with compatible filter sets
- Kit is suitable for use with fluorescence microscopes or high content analysis (HCA) fluorescence multiwall plate scanners
- Easily adaptable for use other fluorescence detection systems
- Assay principles are general and applicable to most eukaryotic cell types, including adherent and non-adherent cells
Specifications
Specifications
| Color | Green, Deep Red |
| Content And Storage | • 1 vial of Calcein AM (live cell indicator), 40 μL, provided as a 2000X (4 mM) solution in DMSO • 1 vial of SYTOX Deep Red Nucleic Acid Stain (dead cell indicator), 40 μL, provided as a 2000X (500 μM) solution in DMSO Store at ≤-20°C, upright in dry place. Protect from light. |
| Description | LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope, High Content Analysis Instrument, Other Fluorescence Detection Systems |
| Product Type | M25 |
| Format | Kit |
| Indicator | Calcein, AM (live cell indicator), SYTOX™ Deep Red Nucleic Acid Stain (dead cell indicator) |
| Product Line | LIVE/DEAD |
| Show More |
Frequently Asked Questions (FAQs)
We have not validated the use of LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red)(Cat. No. L32250) on bacteria, fungi or protozoa. In general, calcein, AM does not work well with bacteria due to their low intracellular esterase activity. Some protozoa may readily engulf cell impermeant stains and result in intracellular staining of live cells.
If an expiration date or warranty period is not posted on any public-facing document, the LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) (Cat. Nos. L32250) have the standard warranty period of 1 year from the date of shipment.
The formulation/identity of components in the solution LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) (Cat. No. L32250) is proprietary.
The concentrations of Calcein, AM and SYTOX Deep Red Nucleic Acid stain in the LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) (Cat. No. L32250) are proprietary. If you need to use a defined concentration of Calcein, AM we offer this as a standalone product (Cat. Nos. C3099 and C3100MP). If you need to use a defined concentration of SYTOX Deep Red Nucleic Acid stain we offer this as a standalone product (Cat. Nos. S11380 and S11380).