missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Dead Cell Stain Sampler Kit

Catalog No. 501137444
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137444 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137444 Supplier Molecular Probes™ Supplier No. L34960

LIVE/DEAD™ Fixable dead cell stains are used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation
  • The stains in the sampler kit have been optimized and validated for use in flow cytometry
  • Stains in this sampler kit have been conveniently packaged in eight 40-test vials to help ensure the stability and performance of the dye over time
  • Amine-reactive dyes in solution will lose their effectiveness over a short period of time
  • It is recommended to use the vial completely once rehydrated or aliquot the solution and store at -80°C, avoiding freeze thaw cycles
  • LIVE/DEAD™ Fixable dead cell stains are available in a wide variety of colors to meet multi-color panel needs
  • The sampler kit provides one vial (40 assays) of each of the eight dead cell stains available

Optimal brightness

  • The LIVE/DEAD™ Fixable dead cell stains were selected based on their fluorescent properties to provide a bright signal when excited with the optimal lasers
  • Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, these dead cell stains are an ideal choice for multi-color experiments

Find the LIVE/DEAD™ Fixable Dead Cell Stain that fits various needs:

  • For UV lasers: blue
  • For violet lasers: aqua, violet, or yellow
  • For blue lasers: green or red
  • For red lasers: far-red or near-IR

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Various
Content And Storage Contians 8 vials (1 vial per color) of the LIVE/DEAD™ fixable dead cell stains and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Dead Cell Stain Sampler Kit
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain Sampler Kit
Dye Type Other Label(s) or Dye(s)
Excitation Wavelength Range 350/416/367/400/495/595/650/750 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD, Molecular Probes
Quantity 1 kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength Range 450/451/526/575/520/615/665/775
Show More Show Less
I need to use a dead cell control for my viability assay. Do you have a protocol for killing cells for this?

Heat killing is commonly used. Place your cells in a tube in buffer and heat at 60oC for 20 minutes. You can also kill your cells by fixing them with ice cold 70% ethanol for 15 minutes. The ethanol-killed cells can then be stored at -20oC until needed, at which point you wash out the ethanol and replace with buffer.

Regarding the LIVE/DEAD Fixable Dead Cell Stain Kits, which can discriminate between live and dead cells using flow cytometry with one emission wavelength. Can these kits be used with microscopy?

This dye gives a dim surface label for live cells, but is internalized and gives a brighter signal for dead cells. Flow cytometry is a very sensitive technique and can easily distinguish between the two populations. Microscopy is not as sensitive and may not be able to distinguish the cells because of a less sensitive detector.

For Research Use Only. Not for use in diagnostic procedures.