missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 99%

Catalog No. AC301221000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC301221000 100 g
AC301220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC301221000 Supplier Thermo Scientific Chemicals Supplier No. 301221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1907-33-1 | C4H9LiO | 80.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1907-33-1
Molecular Formula C4H9LiO
Molecular Weight (g/mol) 80.06
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
PubChem CID 23664764
IUPAC Name lithium;2-methylpropan-2-olate
SMILES [Li+].CC(C)(C)[O-]

Specifications

Color White to Yellow
Density 0.8900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula LiOCH(CH3)3
Quantity 100 g
UN Number 2925
Specific Gravity 0.89
Sublimation Point 283 °C
Solubility Information Solubility in water: vigorous reaction
Formula Weight 80.06
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Lithium tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Harmful if swallowed.
Reacts violently with water.
Precautionary Statement GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if
EINECSNumber 217-611-5

RUO – Research Use Only