missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Standard, 3000 mEq/L (3 N), Ricca Chemical
$414.93 - $414.93
Chemical Identifiers
| CAS | 7447-41-8 |
|---|---|
| Molecular Formula | ClLi |
| Molecular Weight (g/mol) | 42.39 |
| MDL Number | MFCD00011078 |
| InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
| PubChem CID | 433294 |
| ChEBI | CHEBI:48607 |
| IUPAC Name | lithium(1+) chloride |
| SMILES | [Li+].[Cl-] |
Chemical Identifiers
| 7447-41-8 | |
| 42.39 | |
| KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
| CHEBI:48607 | |
| [Li+].[Cl-] |
| ClLi | |
| MFCD00011078 | |
| 433294 | |
| lithium(1+) chloride |
Specifications
| Natural Poly Bottle | |
| 87.0,11.28 | |
| 90.0,11.74 | |
| ClLi | |
| Colorless | |
| Approximately 0°C | |
| [Li+].[Cl-] | |
| 42.39 | |
| 433294 | |
| Lithium Standard |