CAS RN 7447-41-8
Lithium Chloride (Certified), Fisher Chemical™
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
Lithium chloride, ACS, 99% min
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
Electrolyte Solution, Nonaqueous, 2 M LiCl in Ethanol, MilliporeSigma™ Supelco™
Filling solution for reference electrodes used for titrations in nonaqueous solutions.
Lithium chloride, anhydrous, 99%
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 SMILES: [Li+].[Cl-]
Lithium Chloride Solution, 1 M in Ethanol, MilliporeSigma™ Supelco™
Lithium chloride solution is generally used as a liquid desiccant for hybrid solar desiccant cooling systems. Though it is corrosive in nature, it does not vaporize in air at ambient conditions.
Lithium chloride, anhydrous, 98+%
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium;chloride SMILES: [Li+].[Cl-]
Lithium chloride, 99+%, ACS reagent
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 SMILES: [Li+].[Cl-]
MilliporeSigma™ Lithium Chloride 8M, OmniPur™, Calbiochem™,
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
Lithium chloride, ultra dry, 99.995% (metals basis)
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 SMILES: [Li+].[Cl-]
Lithium chloride, 1M aq. soln.
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 SMILES: [Li+].[Cl-]
Lithium chloride, 99%, For Analysis, Anhydrous
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
Lithium chloride, anhydrous, 99%, Thermo Scientific™
Thermo Scientific Chemicals Lithium chloride, 99%, for molecular biology
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium;chloride SMILES: [Li+].[Cl-]
Lithium chloride, 99%, extra pure
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M Synonym: lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
Lithium Standard, 3000 mEq/L (3 N), Ricca Chemical
CAS: 7447-41-8 Molecular Formula: ClLi Molecular Weight (g/mol): 42.39 MDL Number: MFCD00011078 InChI Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M PubChem CID: 433294 ChEBI: CHEBI:48607 IUPAC Name: lithium(1+) chloride SMILES: [Li+].[Cl-]
- 1
- 2