missing translation for 'onlineSavingsMsg'
Learn More

Lithium, plasma standard solution, Specpure™ Li 1000μg/mL

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

$97.97 - $328.34

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13821AD
View Documents
Thermo Scientific Chemicals
013821AD
50 mL
Each for $97.97
Only null left
 
AA13821AE
View Documents
Thermo Scientific Chemicals
013821AE
100 mL
Each for $147.23
Only null left
 
AA13821AP
View Documents
Thermo Scientific Chemicals
013821AP
500 mL
Each for $328.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

554-13-2
73.89
XGZVUEUWXADBQD-UHFFFAOYSA-L
CHEBI:6504
CLi2O3
MFCD00011084
11125
[Li+].[Li+].[O-]C([O-])=O

Specifications

554-13-2 , 7697-37-2 , 7732-18-5
Liquid
MFCD00011084
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium;carbonate
50 mL
CHEBI:6504
Lithium
23 hPa (17mm Hg) at 20°C
CLi2O3
Colorless
[Li+].[Li+].[O-]C([O-])=O
73.89
11125
Nitric acid: 5.0%; Lithium carbonate: 0.53%; Water: 94.47%
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only