missing translation for 'onlineSavingsMsg'
Learn More

Lithium Nitrate, Reagent Grade, Ricca Chemical

Catalog No. RDCL0300500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCL0300500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCL0300500 Supplier Ricca Chemical Company Supplier No. RDCL0300500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General laboratory reagent grade lithium nitrate, supplied as a solid crystalline powder.

  • Chemical Grade: Reagent
  • Physical State: Solid
  • Appearance: Colorless/white
  • Molecular Formula: LiNO3
  • Molecular Weight: 68.94
  • Boiling Point: 600°C (decomposes)
  • Relative Density: 2.38
  • pH: Neutral (aqueous solution)
  • Hazard Classification: Oxidizer (Class 5.1)

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium(1+) nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Assay Percent Range 98.00%
Linear Formula

LiNO3

InChI Formula InChI=1S/Li.NO3/c2-1(3)4/q+1;-1
Formula Weight 68.94
Percent Purity ≥98%
Grade Reagent
Manufacturing Origin USA
Packaging White Poly Wide Mouth Bottle
Appearance White Crystalline Powder
Chemical Name or Material Lithium Nitrate
Show More Show Less
ShelfLife 60 months