missing translation for 'onlineSavingsMsg'
Learn More

Lithium manganese(III,IV) oxide, 99.5% (metals basis)

Catalog No. AA4025014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4025014 25 g
AA4025022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4025014 Supplier Thermo Scientific Chemicals Supplier No. 04025014
Only null left

CAS: 12057-17-9 | LiMn2O4 | 180.81 g/mol

It is used as anode materials of high-performance lithium ion battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12057-17-9
Molecular Formula LiMn2O4
Molecular Weight (g/mol) 180.81
MDL Number MFCD01114233
InChI Key KEZPOJYZIWMPGY-UHFFFAOYSA-N

Specifications

Melting Point >400°C
Physical Form Powder
Quantity 25 g
Linear Formula LiMn2O4
Solubility Information Insoluble in water.
Formula Weight 180.81
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lithium manganese(III,IV) oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only