missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Lithium-Ion Battery
Description
- Accessory
- For use with HandyStep™ Touch and Touch S Electronic Repeating Pipette
Specifications
Specifications
| Battery Type | Lithium-ion |
| Description | Replacement lithium-ion battery |
| Color | Black/White |
| For Use With | BRAND HandyStep Touch/Touch S |
| Material | Plastic |
| Type | Battery |
| For Use With (Equipment) | HandyStep Touch / Touch S |
| Quantity | 1 |