missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide hydrate, Puratronic™, 99.995% (metals basis)

Catalog No. AA1073814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1073814 25 g
AA1073806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1073814 Supplier Thermo Scientific Chemicals Supplier No. 01073814
Only null left

CAS: 85017-80-7 | ILi | 133.84 g/mol

Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85017-80-7
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00149769
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci
PubChem CID 23688290
IUPAC Name lithium;iodide;hydrate
SMILES [Li+].[I-]

Specifications

Melting Point 73°C (in -H2O)
Physical Form Crystalline Aggregates
Quantity 25 g
Merck Index 14,5535
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 133.84 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Sensitivity Air and light sensitive; Hygroscopic
Assay (metals basis)
Chemical Name or Material Lithium iodide hydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-822-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only