missing translation for 'onlineSavingsMsg'
Learn More

Lithium ICP Standard, 1000 ppm Li in 3% HNO3, Ricca Chemical

Catalog No. PLI1KN100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLI1KN100 100 mL Natural LDPE Bottle
PLI1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLI1KN100 Supplier Ricca Chemical Company Supplier No. PLI1KN100
Only null left

Chemical Identifiers

CAS 554-13-2 , 7697-37-2 , 554-13-2 , 7732-18-5
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Li
CAS Min % 92.0, 5.4, 0.49
CAS Max % 96.0, 5.62, 0.51
Equivalence 1 mL = 1 mg Li
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Lithium ICP Standard
Show More Show Less