missing translation for 'onlineSavingsMsg'
Learn More

Lithium ICP Standard, 10,000 ppm Li in 5% HNO3, Ricca Chemical

$539.40 - $539.40

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLI10KN-100
SDS View Documents
Ricca Chemical Company
PLI10KN100
100 mL Natural LDPE Bottle
Each for $539.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

554-13-2
73.89
XGZVUEUWXADBQD-UHFFFAOYSA-L
CHEBI:6504
[Li+].[Li+].[O-]C([O-])=O
CLi2O3
MFCD00011084
11125
dilithium(1+) carbonate

Specifications

Natural LDPE Bottle
10,000ppm Li
81.0,17.08,4.29
Liquid
MFCD00011084
< 2
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium(1+) carbonate
100 mL
CHEBI:6504
554-13-2 , 7697-37-2 , 554-13-2 , 7732-18-5
77.0,16.41,4.13
1 mL = 10 mg Li
CLi2O3
Colorless
Standard
[Li+].[Li+].[O-]C([O-])=O
73.89
11125
Lithium ICP Standard
Videos
SDS
Documents

Documents