missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, 13.30 M, Ricca Chemical

Catalog No. R4373000500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4373000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4373000500 Supplier Ricca Chemical Company Supplier No. R4373000500A

Chemical Identifiers

CAS 7447-41-8 , 7732-18-5
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Color Colorless
CAS Min % 63.0, 35.35
CAS Max % 65.0, 36.79
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 6 to 8
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 13.30Molar
Chemical Name or Material Lithium Chloride
Show More Show Less