missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonate de lithium 98,0+%, TCI America™
Description
- Chemical Name: Lithium Carbonate
- Formula: Li2CO3
- Molecular Weight: 73.89
- Appearance: White powder to crystal
- Purity: ≥98.0% (neutralization titration)
- Melting Point: 723 °C
- Solubility: Slightly soluble in water
- Applications: Battery research, inorganic chemistry, and synthetic reagents
- Storage: Room temperature; store in a cool, dark place (<15°C)
- Notes: For laboratory research purposes only; not for drug or household use
Spécifications
Spécifications
| Melting Point | 723°C |
| Color | Blanc |
| Physical Form | Poudre |
| Quantity | 25 g |
| Assay Percent Range | 98.00% |
| Linear Formula | Li2CO3 |
| Merck Index | 14, 5527 |
| Solubility Information | Slightly soluble in water |
| Formula Weight | 73.89 |
| Percent Purity | ≥98.0% (T) |
| Afficher plus de résultats |
Sécurité et manipulation
| EINECSNumber | (1)-0154 |
| RTECSNumber | OJ5800000 |
| TSCA | Yes |