missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bis(pentafluoroethanesulfonyl)imide 98.0+%, TCI America™

Numéro de catalogue. L02675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
L02675G 5 g
L02671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. L02675G Fournisseur TCI America Code fournisseur L02675G
Il en reste null

Identifiants chimiques

CAS 132843-44-8
Molecular Formula C4F10LiNO4S2
Molecular Weight (g/mol) 387.09
MDL Number MFCD22374096
InChI Key ACFSQHQYDZIPRL-UHFFFAOYSA-N
Synonym Bis(pentafluoroethanesulfonyl)imide Lithium Salt
PubChem CID 12096715
IUPAC Name lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane
SMILES [Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F

Spécifications

Melting Point 328°C
Color White
Quantity 5 g
UN Number 2923
Formula Weight 387.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Bis(pentafluoroethanesulfonyl)imide
EINECSNumber (2)-3839
TSCA Yes