missing translation for 'onlineSavingsMsg'
Learn More

Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane

Catalog No. AAL08269AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08269AP 500 mL
AAL08269AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08269AP Supplier Thermo Scientific Chemicals Supplier No. L08269AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15405-86-4 | C32H72AlLiN4 | 546.878 g/mol

Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.

Chemical Identifiers

CAS 15405-86-4
Molecular Formula C32H72AlLiN4
Molecular Weight (g/mol) 546.878
MDL Number MFCD00051918
InChI Key ISBVOXPIPNIBOM-UHFFFAOYSA-N
Synonym lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
PubChem CID 57369578
IUPAC Name lithium;tetrakis(dibutylamino)alumanuide
SMILES [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC

Specifications

Melting Point -54°C
Density 0.86
Linear Formula LiAl[N(CH2CH2CH2CH3)2]4
Quantity 500 mL
UN Number UN1993
Sensitivity Air and moisture sensitive
Formula Weight 546.87
Chemical Name or Material Lithium Aluminum Di-n-butylamide
Hazard Category H225-H315-H319-H332-H335-H360FD
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only