Learn More
Lithium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Li), LabChem™
Supplier: LabChem LC161801
Specifications
| Lithium AA Standard | |
| Poly Bottle | |
| 95.9,3.1,1 | |
| 1g/mL | |
| LiNO3 | |
| LiNO3 | |
| UN2031 | |
| Soluble in water | |
| [Li+].[O-][N+]([O-])=O | |
| 68.94 | |
| 10129889 | |
| 68.96 |
| 1000ppm (1mL = 1mg Li) | |
| 7790-69-4 , 7697-37-2 , 7790-69-4 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Li) | |
| MFCD00011094 | |
| Colorless | |
| IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
| lithium(1+) nitrate | |
| 500 mL | |
| CHEBI:63315 | |
| Certified |
Chemical Identifiers
| 7790-69-4 | |
| 68.94 | |
| IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
| CHEBI:63315 | |
| [Li+].[O-][N+]([O-])=O |
| LiNO3 | |
| MFCD00011094 | |
| 10129889 | |
| lithium(1+) nitrate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature