Learn More
Gilson™ Lite Pipette Service
Catalog No. SVC101102G
Change view
Click to view available options
Includes:
EasyCheck Multi
EasyCheck Single
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
SVC101102G EasyCheck Multi
SVC101101G EasyCheck Single
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SVC101102G Supplier Gilson™ Supplier No. SVC101102
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lite pipette service is ideal for labs that need dependable preventative maintenance (PM). The standard PM will be performed on each pipette along with a gravimetric check, ensuring the performance of pipettes.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • 5 day turn around time
Specifications
For Use With (Equipment) Pipettes
Description Easy Check Multi
Duration 5 Days
Product Type Service
Includes EasyCheck Multi