missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIS-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Zebrafish, X. laevis, X. tropicalis, Chicken, Bovine, Pig, Cat, Orangutan and Chimpanzee
This locus was identified as encoding a gene that when mutated or lost caused the lissencephaly associated with Miller-Dieker lissencephaly syndrome. This gene encodes the non-catalytic alpha subunit of the intracellular Ib isoform of platelet-activating factor acteylhydrolase, a heterotrimeric enzyme that specifically catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist: one composed of multiple subunits, the other, a single subunit. In addition, a single-subunit isoform of this enzyme is found in serum.
Spécifications
Spécifications
| Antigène | LIS-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugué | Unconjugated |
| Formule | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gène | PAFAH1B1 |
| Numéro d’acquisition du gène | P43034, P63004, P63005 |
| Alias du gène | RP23-194P5.2, LIS-1, Lis1, MGC25297, MMS10-U, Mdsh, Ms10u, Pafaha |
| Symboles de gène | Pafah1b1 |
| Afficher plus de résultats |