missing translation for 'onlineSavingsMsg'
Learn More

Liquefied Phenol, USP, 89%, Spectrum™ Chemical

Catalog No. 18603913
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603913 200 L Poly-Coated Glass Bottle
18603911 500 mL Poly-Coated Glass Bottle
18603912 6 x 500 mL Poly-Coated Glass Bottle
18603908 4 L Poly-Coated Glass Bottle
18603910 4 x 4 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603913 Supplier Spectrum Chemical Mfg Cor Supplier No. PH125200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PH125, 108-95-2, C6H6O

Spectrum™ Chemical Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 108-95-2 , 7732-18-5
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 89%, 11%
Packaging Poly-Coated Glass Bottle
Quantity 200 L
UN Number UN2821
Percent Purity 89%
Grade USP
Physical Form Liquid