missing translation for 'onlineSavingsMsg'
Learn More

Linolenic Acid 70.0+%, TCI America™

Catalog No. L005025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L005025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L005025ML Supplier TCI America Supplier No. L005025ML

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18H30O2
Molecular Weight (g/mol) 278.436
MDL Number MFCD00065720
InChI Key DTOSIQBPPRVQHS-PDBXOOCHSA-N
Synonym linolenic acid, alpha-linolenic acid, linolenate, 9z,12z,15z-octadeca-9,12,15-trienoic acid, a-linolenic acid, cis,cis,cis-9,12,15-octadecatrienoic acid, all-cis-9,12,15-octadecatrienoic acid, alpha-linolenate, 9-cis,12-cis,15-cis-octadecatrienoic acid, z,z,z-9,12,15-octadecatrienoic acid
PubChem CID 5280934
ChEBI CHEBI:27432
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -11°C
Color Yellow
Boiling Point 232°C
Quantity 25 mL
Formula Weight 278.44
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Linolenic Acid
EINECSNumber (2)-0609
TSCA Yes
Recommended Storage Refrigerator