missing translation for 'onlineSavingsMsg'
Learn More

Linolenic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014230
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014230 5 mL
111014229 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014230 Supplier Merck Emd Millipore Supplier No. 621605ML
Featured Industries: Food and Beverages

Specifications

CAS 463-40-1
Density 0.914 g/mL (at 25°C)
Flash Point 113°C - closed cup
Physical Form Neat
Linear Formula CH3(CH2 CH=CH)3(CH2)7 CO2 H
MDL Number MFCD00065720
Melting Point -11°C
Boiling Point 230°C to 232°C (1 mmHg)
Synonym alpha-Lnn; cis,cis,cis-9,12,15-Octadecatrienoic acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 278.43
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less