missing translation for 'onlineSavingsMsg'
Learn More

Linoleic Acid 97.0+%, TCI America™

Catalog No. L01245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01245G 5 g
L012425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01245G Supplier TCI America Supplier No. L01245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 177°C
Quantity 5 g
Formula Weight 280.45
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Linoleic Acid
EINECSNumber (2)-0609
RTECSNumber RF9990000
TSCA Yes
Recommended Storage Refrigerator