missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linoleic acid, 99%

Catalog No. AC215041000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215041000 100 mL Glass bottle
AC215040050 5 mL Glass bottle
AC215040250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215041000 Supplier Thermo Scientific Chemicals Supplier No. 215041000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.45
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -5.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 229.0°C to 230.0°C (16.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4687 to 1.4707
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Quantity 100 mL
Beilstein 02,IV,1754
Specific Gravity 0.9
Merck Index 15,556
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether
Formula Weight 280.45
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Linoleic acid
Show More Show Less
Hazard Category Warning
EINECSNumber 200-470-9

RUO – Research Use Only