Learn More
Eisco™ Linear Expansion Apparatus
Catalog No. S23742
Change view
Click to view available options
Grade:
9 to Undergraduate
Catalog No. Grade
S23742 9 to Undergraduate
1 options
Catalog No. S23742 Supplier Eisco™ Supplier No. PH0371A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Linear Expansion Apparatus is fitted on heavy channel base with adjusting screw and micrometer screw on opposite ends.
  • Consists of metal rod 500 mm L, 4.5 mm dia.
  • Rod enclosed in well-lagged metal tube with 3 tubules for steam inlet, outlet, thermometer.
  • Rod to be investigated supported in steam jacket by detachable rubber bungs at tube ends.
  • Micrometer screw reading to 0.01 mm for measuring expansion.
  • Supplied with one each of aluminum, brass, iron expansion rods, without thermometer.
Specifications
Grade 9 to Undergraduate
Includes 1 Base, 1 Dial Gauge, 3 Metal Rods
Length (English) 27 in.
Length (Metric) 68.6 cm
Product Type Linear Expansion Apparatus
Description Adjusting screw and micrometer screw on opposite ends