missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ligustrazine hydrochloride

Catalog No. AC467640010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467640010 1 g
AC467640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467640010 Supplier Thermo Scientific Chemicals Supplier No. 467640010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76494-51-4
Molecular Formula C8H12N2·HCl

Specifications

Quantity 1 g
Boiling Point 193.0°C (730.0 mmHg)
Formula Weight 172.66
Chemical Name or Material Ligustrazine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only