accessibility menu, dialog, popup

UserName

CAS RN 76494-51-4

IUPAC Name: tetramethylpyrazine hydrochloride
Synonyms: tetramethylpyrazine hydrochloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: RQKFOGXUTRDQPB-UHFFFAOYSA-N
SMILES: Cl.CC1=NC(C)=C(C)N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results