missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine 99.0+%, TCI America™

Supplier:  TCI America L0156100G

 View more versions of this product

Catalog No. L0156100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lidocaine
137-58-6
182°C
Crystal-Powder at 20°C
MFCD00026733
NNJVILVZKWQKPM-UHFFFAOYSA-N
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
3676
234.34
69°C
White
100 g
C14H22N2O
lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C
234.343
CHEBI:6456
≥99.0% (HPLC,T)

Chemical Identifiers

137-58-6
234.343
NNJVILVZKWQKPM-UHFFFAOYSA-N
3676
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
C14H22N2O
MFCD00026733
lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
CHEBI:6456
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0273&(9)-1510

RTECSNumber : AN7525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions