missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine 99.0+%, TCI America™

Catalog No. p-7131846
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L015625G 25 g
L0156100G 100 g
L0156500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L015625G Supplier TCI America Supplier No. L015625G

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
MDL Number MFCD00026733
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
Synonym lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
PubChem CID 3676
ChEBI CHEBI:6456
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)NC1=C(C=CC=C1C)C

Specifications

Melting Point 69°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 182°C
Formula Weight 234.34
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material Lidocaine
EINECSNumber (3)-0273&(9)-1510
RTECSNumber AN7525000
TSCA Yes