accessibility menu, dialog, popup

UserName

H3CNH3CONHCH3H3C

CAS RN 137-58-6

IUPAC Name: 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
Synonyms: lidocaine xylocaine 5% spinal after burn spray zilactin-L L-caine octocaine-100 after burn gel leostesin zingo duncaine
Molecular Weight (g/mol): 234.34
Molecular Formula: C14H22N2O
InChi Key: NNJVILVZKWQKPM-UHFFFAOYSA-N
SMILES: CCN(CC)CC(=O)NC1=C(C)C=CC=C1C
Grade 
  • (1)
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)