missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine N-ethyl bromide, MilliporeSigma™ Supelco™

$125.59 - $467.74

Specifications

CAS 21306-56-9
Physical Form Neat
Linear Formula C16H27 BrN2 O
MDL Number MFCD00083182
Synonym N-(2,6-Dimethylphenylcarbamoylmethyl)triethylammonium bromide; QX-314
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013538
View Documents
Merck Emd Millipore
L578350MG
50 mg
Each for $125.59
Only null left
 
111013537
View Documents
Merck Emd Millipore
L5783250MG
250 mg
Each for $467.74
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

21306-56-9
C16H27 BrN2 O
N-(2,6-Dimethylphenylcarbamoylmethyl)triethylammonium bromide; QX-314
Drug Analysis
Analytical Standard
Neat
MFCD00083182
Analytical Standard
343.30
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'