missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine hydrochloride monohydrate, 98%

Catalog No. AAJ6303514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6303514 25 g
AAJ6303506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6303514 Supplier Thermo Scientific Chemicals Supplier No. J6303514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6108-05-0 | C14H25ClN2O2 | 288.82 g/mol

It is also used as a fast voltage-gated sodium channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also used as a fast voltage-gated sodium channel blocker.

Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14H25ClN2O2
Molecular Weight (g/mol) 288.82
MDL Number MFCD00150329
InChI Key YECIFGHRMFEPJK-UHFFFAOYSA-N
Synonym lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
PubChem CID 16219577
ChEBI CHEBI:60791
IUPAC Name {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
SMILES O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

Melting Point 75°C to 79°C
Color White
Quantity 25 g
UN Number UN2811
Merck Index 14,5482
Solubility Information Soluble in water (50mg/ml),chloroform,ethanol,and benzene. Insoluble in diethyl ether.
Formula Weight 288.82 (270.80 Anhydrous)
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Lidocaine hydrochloride monohydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber AN7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only