missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ LiChrosorb™ RP-18 HPLC Column, 5 μm Particle Size

LiChrosorb™ RP-18 HPLC Columns are classified as L1 USP columns and are primarily used for reversed-phase HPLC of neutral, acidic and weak basic compounds. The totally porous irregular particles ensure reliable results.

$1637.71 - $1780.24

Specifications

Particle Size 5 μm
Pore Size 100 Å
Surface Area 300 m2/g
Stationary Phase C18 (octadecyl)
Product Line LiChrosorb
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
111003364
View Documents
MilliporeSigma™ Supelco™
54951
15 cm 4.6 mm
Each for $1,637.71
Only null left
 
111003264
View Documents
MilliporeSigma™ Supelco™
54949
25 cm 4.6 mm
Each for $1,780.24
Only null left
 
Description

Description

LiChrosorb™ RP-18 HPLC column comprises of totally porous irregular particles made of silica with an octadecyl derivative. This highly reliable sorbent is suitable for reversed-phase HPLC of neutral, acidic and weak basic compounds, according to L1 USP specifications.
Specifications

Specifications

5 μm
300 m2/g
LiChrosorb
No
HPLC Column
100 Å
C18 (octadecyl)
C18 (octadecyl) Phase/Silica Particle Platform
Reversed Phase
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

LiChrosorb is a registered trademark of Merck KGaA, Darmstadt, Germany