missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium octyl sulfate , ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013090
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013090 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013090 Supplier Merck Emd Millipore Supplier No. 7507310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00007470
Synonym Octyl sulfate sodium salt

Specifications

Melting Point 195°C
CAS 142-31-4
Linear Formula CH3(CH2)7 OSO3 Na
Quantity 10 g
Formula Weight 232.27
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Line LiChropur
RTECSNumber WT1175000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany