missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium Formate Solution, MilliporeSigma™ Supelco™

Catalog No. 111022627
Change view
Click to view available options
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022627 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022627 Supplier Merck Emd Millipore Supplier No. 51197100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium formate solution is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS).

  • Extensive QC testing ensures the highest specifications
  • Optimized to improve ionization and resolution
  • Extremely low levels of inorganic and organic impurities
  • Manufactured specifically for accurate and fast LC-MS

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
IUPAC Name sodium formate
SMILES [Na+].[O-]C=O

Specifications

Quantity 100 mL
Linear Formula HCOONa
Formula Weight 68.01
Concentration or Composition (by Analyte or Components) 10 mM NaOH in isopropanol/water 1:1 (+0.2% HCOOH)

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany