missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-octanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 5324-84-5, 207596-29-0 |
| Melting Point | ≥300°C |
| Linear Formula | CH3(CH2)6 CH2 SO3 Na · H2O |
| Quantity | 50 g |
| Molecular Weight (g/mol) | 216.28 |
| Formula Weight | 234.29 |
| Percent Purity | ≥99.0% (T) |
| Grade | For ion pair chromatography |
| Physical Form | Powder |
| Product Line | LiChropur |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany