missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-N-trimethylsilyltrifluoroacetamide Activated I, For GC Derivatization, MilliporeSigma™ Supelco™
Description
Derivatizing agent for the trimethylsilylation of -OH and -NH groups for GC and GC-MS
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-(trimethylsilyl)trifluoroacetamide |
| CAS | 24589-78-4 |
| Packaging | Clear Glass Bottle |
| Refractive Index | n20/D 1.380 |
| Quantity | 5 mL |
| Synonym | MSTFA activated I |
| Molecular Weight (g/mol) | 199.25 |
| Grade | For GC derivatization |
| Physical Form | Liquid |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany