missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1 trimethylchlorosilane, ≥97%, MilliporeSigma™ Supelco™

Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).

$105.99 - $902.68

Specifications

CAS 24589-78-4
Product Line LiChropur
Flash Point 26°C - closed cup
Linear Formula C6H12F3 NOSi
MDL Number MFCD00000411
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111020206
View Documents
Merck Emd Millipore
694781MLF
1 mL
Each for $105.99
Only null left
 
111018572
View Documents
Merck Emd Millipore
694785MLF
5 mL
Each for $421.23
Only null left
 
111018139
View Documents
Merck Emd Millipore
6947810X1MLF
10 x 1 mL
Each for $902.68
Only null left
 
Description

Description

Specifications

Specifications

24589-78-4
26°C - closed cup
MFCD00000411
199.25
GC Derivatization
LiChropur
C6H12F3 NOSi
Organic Standard
≥97%
Videos
SDS
Documents

Documents

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany