missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1 trimethylchlorosilane, ≥97%, MilliporeSigma™ Supelco™

Catalog No. 111020206
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020206 1 mL
111018572 5 mL
111018139 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020206 Supplier Merck Emd Millipore Supplier No. 694781MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).

Specifications

Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
CAS 24589-78-4
Product Line LiChropur
Flash Point 26°C - closed cup
Concentration 1%
Linear Formula C6H12F3 NOSi
Technique Gas Chromatography
MDL Number MFCD00000411
Type Organic Standard
Solvent or Matrix Neat
Molecular Weight (g/mol) 199.25
Quantity 1 mL
Formula Weight 199.25
Percent Purity ≥97%
Grade GC Derivatization
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany